missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2-bromoisobutyrate, 97%

Catalog No. p-7044784
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0821609 10 g
AAL0821618 50 g
AAL0821630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0821609 Supplier Thermo Scientific Chemicals Supplier No. L0821609
Only null left

CAS: 23877-12-5 | C8H15BrO2 | 223.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23877-12-5
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD00051908
InChI Key IGVNJALYNQVQIT-UHFFFAOYSA-N
PubChem CID 90290
IUPAC Name tert-butyl 2-bromo-2-methylpropanoate
SMILES CC(C)(C)OC(=O)C(C)(C)Br

Specifications

Density 1.18
Boiling Point 35°C to 37°C (1 mmHg)
Refractive Index 1.436
Quantity 10 g
Beilstein 1905654
Synonym tert-butyl 2-bromoisobutyrate, t-butyl 2-bromo isobutyrate, 2-bromoisobutyric acid tert-butylester, tert-butyl 2-bromo-2-methylpropionate, tert-butyl alpha-bromoisobutyrate, t-butyl 2-bromoisobutylate, t-butyl 2-bromoisobutyrate, 2-bromoisobutyric acid tert-butyl ester, propanoic acid, 2-bromo-2-methyl-, 1,1-dimethylethyl ester, t-butyl2-bromoisobutyrate
Formula Weight 223.12
Percent Purity 97%
Chemical Name or Material tert-Butyl 2-bromoisobutyrate
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 245-924-7
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only