Learn More
tert-Butanol, Analytical Standard, ≥99.8% (GC), MilliporeSigma™ Supelco™
tert-butanol can find applications as an ideal reaction medium or solvent for the lipase-catalyzed biodiesel production.
Supplier:  Merck Emd Millipore 506215MLF
 View more versions of this product
Catalog No. 111019168
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
75-65-0
2.5 (vs. air)
Neat
MFCD00004464
83°C
5 mL
≥99.8% (GC)
31 mmHg (20°C)
0.775 g/mL (at 25°C)
(CH3)3 COH
23°C to 26°C
2-Methyl-2-propanol; tert-Butyl alcohol; Trimethyl carbinol
74.12
Analytical Standard
Product Suggestions
Product Suggestions