missing translation for 'onlineSavingsMsg'
Learn More

tert-Butanol, Analytical Standard, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7193455
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020592 1 mL
111019168 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020592 Supplier Merck Emd Millipore Supplier No. 506211MLF
Only null left

tert-butanol can find applications as an ideal reaction medium or solvent for the lipase-catalyzed biodiesel production.

Specifications

CAS 75-65-0
Vapor Pressure 31 mmHg (20°C)
Vapor Density 2.5 (vs. air)
Density 0.775 g/mL (at 25°C)
Physical Form Neat
Linear Formula (CH3)3 COH
MDL Number MFCD00004464
Melting Point 23°C to 26°C
Boiling Point 83°C
Synonym 2-Methyl-2-propanol; tert-Butyl alcohol; Trimethyl carbinol
Quantity 1 mL
Formula Weight 74.12
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less