missing translation for 'onlineSavingsMsg'
Learn More

tert-Amylbenzene 97.0+%, TCI America™

Catalog No. A075125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A075125ML 25 mL
A0751500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A075125ML Supplier TCI America Supplier No. A075125ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2049-95-8
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00015187
InChI Key QHTJSSMHBLGUHV-UHFFFAOYSA-N
Synonym tert-amylbenzene, tert-pentylbenzene, 2-methyl-2-phenylbutane, 1,1-dimethylpropyl benzene, benzene, 1,1-dimethylpropyl, benzene, tert-pentyl, 2-phenyl-2-methylbutane, t-amylbenzene, 2-methylbutan-2-yl benzene, tert-amyl benzene
PubChem CID 16295
IUPAC Name 2-methylbutan-2-ylbenzene
SMILES CCC(C)(C)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 191°C
Quantity 25 mL
UN Number 3295
Formula Weight 148.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Amylbenzene
EINECSNumber (3)-0022
RTECSNumber DA6720000
TSCA Yes