missing translation for 'onlineSavingsMsg'
Learn More

Terpineol, Practical, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor T1039200LTBL

 View more versions of this product

Catalog No. 18614331

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Terpineol
1
Amber Glass Bottle
200 L
154.24 g/mol
Liquid
8000-41-7
213°C to 224°C
1.482-1.484
0.932 - 0.937
Practical

Chemical Identifiers

8000-41-7
154.24 g/mol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.