accessibility menu, dialog, popup

UserName

CAS RN 8000-41-7

IUPAC Name: 2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
Synonyms: terpineol 1-α-terpineol α-terpineol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: WUOACPNHFRMFPN-UHFFFAOYNA-N
SMILES: CC1=CCC(CC1)C(C)(C)O
Boiling Point 
  • (2)
  • (5)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (5)