missing translation for 'onlineSavingsMsg'
Learn More

Terpineol, Practical, Spectrum™ Chemical

Catalog No. p-7166631
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614329 100 mL Amber Glass Bottle
18614333 500 mL Amber Glass Bottle
18614332 4 L Amber Glass Bottle
18614330 20 L Amber Glass Bottle
18614331 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18614329 Supplier Spectrum Chemical Mfg Cor Supplier No. T1039100ML
Only null left

Chemical Identifiers

CAS 8000-41-7
Molecular Weight (g/mol) 154.24 g/mol

Specifications

CAS Max % 1
Boiling Point 213°C to 224°C
Packaging Amber Glass Bottle
Refractive Index 1.482-1.484
Quantity 100 mL
Specific Gravity 0.932 - 0.937
Grade Practical
Physical Form Liquid
Chemical Name or Material Terpineol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.