missing translation for 'onlineSavingsMsg'
Learn More

Terpineol, mixed isomers, 98%

CAS: 8000-41-7 | C10H18O | 154.25 g/mol

$188.89 - $730.84

Chemical Identifiers

CAS 8000-41-7
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00001557,MFCD00166983
InChI Key WUOACPNHFRMFPN-UHFFFAOYNA-N
Synonym alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
PubChem CID 17100
ChEBI CHEBI:22469
IUPAC Name 2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
SMILES CC1=CCC(CC1)C(C)(C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60146AP
View Documents
Thermo Scientific Chemicals
H60146AP
500 mL
Each for $188.89
Only null left
 
AAH60146K4
View Documents
Thermo Scientific Chemicals
H60146K4
2.5 L
Each for $730.84
Only null left
 
Description

Description

Terpineol is used in perfumes, cosmetics and flavorings. It is also used in the chemical industries and personal-domestic use. It acts as a cleaning and washing agent, disinfectant, food additive and odor agent. It finds applications in fragrance compositions, disinfectants, polishes and various household cleaning products. It is also used as high-grade solvent and deodorizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Terpineol is used in perfumes, cosmetics and flavorings. It is also used in the chemical industries and personal-domestic use. It acts as a cleaning and washing agent, disinfectant, food additive and odor agent. It finds applications in fragrance compositions, disinfectants, polishes and various household cleaning products. It is also used as high-grade solvent and deodorizer.

Solubility
Miscible with organic solvents. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications

Chemical Identifiers

8000-41-7
154.25
WUOACPNHFRMFPN-UHFFFAOYNA-N
17100
2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
C10H18O
MFCD00001557,MFCD00166983
alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
CHEBI:22469
CC1=CCC(CC1)C(C)(C)O

Specifications

8000-41-7
0.934
91°C (196°F)
1.484
500 mL
Miscible with organic solvents. Slightly miscible with water.
CC1=CCC(CC1)C(C)(C)O
154.25
CHEBI:22469
98%
28°C to 36°C
213°C to 218°C
C10H18O
MFCD00001557,MFCD00166983
alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
WUOACPNHFRMFPN-UHFFFAOYNA-N
2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
17100
154.25
Terpineol, mixed isomers
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 232-268-1

RTECSNumber : WZ6600000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only