Learn More
Terpineol, mixed isomers, 98%
CAS: 8000-41-7 | C10H18O | 154.25 g/mol
Supplier: Thermo Scientific Chemicals H60146AP
Description
Terpineol is used in perfumes, cosmetics and flavorings. It is also used in the chemical industries and personal-domestic use. It acts as a cleaning and washing agent, disinfectant, food additive and odor agent. It finds applications in fragrance compositions, disinfectants, polishes and various household cleaning products. It is also used as high-grade solvent and deodorizer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTerpineol is used in perfumes, cosmetics and flavorings. It is also used in the chemical industries and personal-domestic use. It acts as a cleaning and washing agent, disinfectant, food additive and odor agent. It finds applications in fragrance compositions, disinfectants, polishes and various household cleaning products. It is also used as high-grade solvent and deodorizer.
Solubility
Miscible with organic solvents. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications
| Terpineol | |
| 8000-41-7 | |
| 0.934 | |
| 91°C (196°F) | |
| 1.484 | |
| 500 mL | |
| Miscible with organic solvents. Slightly miscible with water. | |
| CC1=CCC(CC1)C(C)(C)O | |
| 154.25 | |
| CHEBI:22469 | |
| 98% |
| mixed isomers | |
| 28°C to 36°C | |
| 213°C to 218°C | |
| C10H18O | |
| MFCD00001557,MFCD00166983 | |
| alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol | |
| WUOACPNHFRMFPN-UHFFFAOYNA-N | |
| 2-(4-methylcyclohex-3-en-1-yl)propan-2-ol | |
| 17100 | |
| 154.25 |
Chemical Identifiers
| 8000-41-7 | |
| 154.25 | |
| WUOACPNHFRMFPN-UHFFFAOYNA-N | |
| 17100 | |
| 2-(4-methylcyclohex-3-en-1-yl)propan-2-ol |
| C10H18O | |
| MFCD00001557,MFCD00166983 | |
| alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol | |
| CHEBI:22469 | |
| CC1=CCC(CC1)C(C)(C)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 232-268-1
RTECSNumber : WZ6600000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only