missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Terpinen-4-ol 93.0+%, TCI America™
Supplier: TCI America T258125G
| Quantity | 25 g |
|---|
Chemical Identifiers
| C10H18O | |
| MFCD00077431 | |
| +-terpinen-4-ol, unii-hzk34ln80n, s-p-menth-1-en-4-ol, hzk34ln80n, s-1-isopropyl-4-methyl-3-cyclohexen-1-ol, 4-terpinenol, d, +-4-terpineol, 4-terpineol, +, 1s-1-isopropyl-4-methylcyclohex-3-en-1-ol, 4-carvomenthenol, + | |
| (1S)-4-methyl-1-propan-2-ylcyclohex-3-en-1-ol |
Specifications
| (+)-Terpinen-4-ol | |
| Yellow | |
| C10H18O | |
| MFCD00077431 | |
| WRYLYDPHFGVWKC-SNVBAGLBSA-N | |
| (1S)-4-methyl-1-propan-2-ylcyclohex-3-en-1-ol | |
| 2724161 | |
| ≥93.0% (GC) |
| 2438-10-0 | |
| 90°C | |
| 25 g | |
| +-terpinen-4-ol, unii-hzk34ln80n, s-p-menth-1-en-4-ol, hzk34ln80n, s-1-isopropyl-4-methyl-3-cyclohexen-1-ol, 4-terpinenol, d, +-4-terpineol, 4-terpineol, +, 1s-1-isopropyl-4-methylcyclohex-3-en-1-ol, 4-carvomenthenol, + | |
| CC1=CCC(CC1)(C(C)C)O | |
| 154.253 | |
| 154.25 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-2331
TSCA : No