missing translation for 'onlineSavingsMsg'
Learn More

(+)-Terpinen-4-ol 93.0+%, TCI America™

Catalog No. p-7137295
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25815G 5 g
T258125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25815G Supplier TCI America Supplier No. T25815G
Only null left

Chemical Identifiers

CAS 2438-10-0
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00077431
InChI Key WRYLYDPHFGVWKC-SNVBAGLBSA-N
Synonym +-terpinen-4-ol, unii-hzk34ln80n, s-p-menth-1-en-4-ol, hzk34ln80n, s-1-isopropyl-4-methyl-3-cyclohexen-1-ol, 4-terpinenol, d, +-4-terpineol, 4-terpineol, +, 1s-1-isopropyl-4-methylcyclohex-3-en-1-ol, 4-carvomenthenol, +
PubChem CID 2724161
IUPAC Name (1S)-4-methyl-1-propan-2-ylcyclohex-3-en-1-ol
SMILES CC1=CCC(CC1)(C(C)C)O

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
Formula Weight 154.25
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material (+)-Terpinen-4-ol
EINECSNumber (3)-2331
TSCA No