accessibility menu, dialog, popup

UserName

CH3H3C(S)HOCH3

CAS RN 2438-10-0

IUPAC Name: (1S)-4-methyl-1-(propan-2-yl)cyclohex-3-en-1-ol
Synonyms: (1S)-1-isopropyl-4-methylcyclohex-3-en-1-ol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: WRYLYDPHFGVWKC-SNVBAGLBSA-N
SMILES: CC(C)[C@]1(O)CCC(C)=CC1
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results