missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ternidazole, 97%

$326.63 - $906.18

Chemical Identifiers

CAS 1077-93-6
Molecular Formula C7H11N3O3
Molecular Weight (g/mol) 185.18
InChI Key DUOHVNSMLSPTMI-UHFFFAOYSA-N
Synonym ternidazole, unii-4n8r018qb0, 1-3-hydroxypropyl-2-methyl-5-nitro-1h-imidazole, 3-2-methyl-5-nitroimidazol-1-yl propan-1-ol, ternidazol, ternidazolum, ternidazole inn, ternidazol inn-spanish, ternidazolum inn-latin
PubChem CID 68944
IUPAC Name 3-(2-methyl-5-nitroimidazol-1-yl)propan-1-ol
SMILES CC1=NC=C(N1CCCO)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC461952500
View Documents Promotion Details
Thermo Scientific™
461952500
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC461950010
View Documents Promotion Details
Thermo Scientific™
461950010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1077-93-6
185.18
ternidazole, unii-4n8r018qb0, 1-3-hydroxypropyl-2-methyl-5-nitro-1h-imidazole, 3-2-methyl-5-nitroimidazol-1-yl propan-1-ol, ternidazol, ternidazolum, ternidazole inn, ternidazol inn-spanish, ternidazolum inn-latin
3-(2-methyl-5-nitroimidazol-1-yl)propan-1-ol
C7H11N3O3
DUOHVNSMLSPTMI-UHFFFAOYSA-N
68944
CC1=NC=C(N1CCCO)[N+](=O)[O-]

Specifications

1077-93-6
C7H11N3O3
DUOHVNSMLSPTMI-UHFFFAOYSA-N
3-(2-methyl-5-nitroimidazol-1-yl)propan-1-ol
68944
97%
96% min. (HPLC)
ternidazole, unii-4n8r018qb0, 1-3-hydroxypropyl-2-methyl-5-nitro-1h-imidazole, 3-2-methyl-5-nitroimidazol-1-yl propan-1-ol, ternidazol, ternidazolum, ternidazole inn, ternidazol inn-spanish, ternidazolum inn-latin
CC1=NC=C(N1CCCO)[N+](=O)[O-]
185.18
185.18
Ternidazole
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-077-5

SDS
Documents

Documents

RUO – Research Use Only