missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Ternidazole, 97%
Supplier: Thermo Scientific™ 461950010
| Quantity | 1g |
|---|
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 1077-93-6 | |
| 185.18 | |
| ternidazole, unii-4n8r018qb0, 1-3-hydroxypropyl-2-methyl-5-nitro-1h-imidazole, 3-2-methyl-5-nitroimidazol-1-yl propan-1-ol, ternidazol, ternidazolum, ternidazole inn, ternidazol inn-spanish, ternidazolum inn-latin | |
| 3-(2-methyl-5-nitroimidazol-1-yl)propan-1-ol |
| C7H11N3O3 | |
| DUOHVNSMLSPTMI-UHFFFAOYSA-N | |
| 68944 | |
| CC1=NC=C(N1CCCO)[N+](=O)[O-] |
Specifications
| Ternidazole | |
| 96% min. (HPLC) | |
| C7H11N3O3 | |
| ternidazole, unii-4n8r018qb0, 1-3-hydroxypropyl-2-methyl-5-nitro-1h-imidazole, 3-2-methyl-5-nitroimidazol-1-yl propan-1-ol, ternidazol, ternidazolum, ternidazole inn, ternidazol inn-spanish, ternidazolum inn-latin | |
| CC1=NC=C(N1CCCO)[N+](=O)[O-] | |
| 185.18 | |
| 185.18 |
| 1077-93-6 | |
| Glass Bottle | |
| 1g | |
| DUOHVNSMLSPTMI-UHFFFAOYSA-N | |
| 3-(2-methyl-5-nitroimidazol-1-yl)propan-1-ol | |
| 68944 | |
| 97% |
Safety and Handling
EINECSNumber : 214-077-5