missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Terfenadine

Catalog No. 394850
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
394850 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 394850 Supplier Tocris Bioscience™ Supplier No. 3948/50

H1 antagonist. Also KV11.1 (hERG) and Kir6 (KATP) channel blocker

Histamine H1 receptor antagonist. Also blocks KV11.1 (hERG) and Kir6 (KATP) channels (IC50 values are 204 nM and 1.2 μM respectively). Inhibits the delayed rectifier K+ current (IKr) in guinea pig ventricular myocytes (IC50 = 50 nM). Activity prolongs QT and induces Torsades de pointes (TdP); cardiotoxic in vivo.

Chemical Identifiers

CAS 50679-08-8
Molecular Formula C32H41NO2

Specifications

Quantity 50 mg
Formula Weight 471.67
Purity >98%
Molecular Weight (g/mol) 471.69
Chemical Name or Material Terfenadine
Recommended Storage Store at +4°C