missing translation for 'onlineSavingsMsg'
Learn More

Terephthaldehyde monodiethylacetal, 97%, stabilized

Catalog No. p-195168
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC294630250 25 g Glass bottle
AC294631000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC294630250 Supplier Thermo Scientific Chemicals Supplier No. 294630250

CAS: 81172-89-6 | C12H16O3 | 208.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 81172-89-6
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.26
MDL Number MFCD00010217
InChI Key HTMXMFARWHNJDW-UHFFFAOYSA-N
Synonym 4-diethoxymethyl benzaldehyde, terephthalaldehyde mono diethyl acetal, benzaldehyde, 4-diethoxymethyl, terephthaldehyde mono diethyl acetal, terephthalaldehyde mono-diethyl acetal, pubchem22621, acmc-209tbs, 4-diethoxymethyl-benzaldehyde, 4-diethoxymethy benzaldehyde, 4-diethyoxymethyl benzaldehyde
PubChem CID 595993
IUPAC Name 4-(diethoxymethyl)benzaldehyde
SMILES CCOC(OCC)C1=CC=C(C=O)C=C1

Specifications

Color Brown to White
Density 1.0400g/mL
Boiling Point 89°C to 93°C (7.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5050 to 1.5075
Linear Formula CHOC6H4CH(OC2H5)2
Quantity 25 g
Specific Gravity 1.04
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in alcohol,chlorinated solvents and,toluene
Stabilizer 0.1 % triethylamine
Formula Weight 208.26
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material Terephthaldehyde monodiethylacetal, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only