missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terephthalaldehyde 98.0+%, TCI America™
Supplier: TCI America T0010100G
Specifications
| Terephthalaldehyde | |
| 115°C | |
| C8H6O2 | |
| 100 g | |
| KUCOHFSKRZZVRO-UHFFFAOYSA-N | |
| terephthalaldehyde | |
| 12173 | |
| ≥98.0% (GC) |
| 623-27-8 | |
| White-Yellow | |
| MFCD00006949 | |
| 1,4-benzenedicarboxaldehyde, p-phthalaldehyde, terephthaldehyde, terephthaldialdehyde, terephthaladehyde, terephthaldicarboxaldehyde, terephthalic aldehyde, 1,4-phthalaldehyde, p-formylbenzaldehyde, terephtaldehyde | |
| C1=CC(=CC=C1C=O)C=O | |
| 134.134 | |
| 134.13 | |
| Crystalline Powder |
Chemical Identifiers
| 623-27-8 | |
| 134.134 | |
| KUCOHFSKRZZVRO-UHFFFAOYSA-N | |
| 12173 | |
| C1=CC(=CC=C1C=O)C=O |
| C8H6O2 | |
| MFCD00006949 | |
| 1,4-benzenedicarboxaldehyde, p-phthalaldehyde, terephthaldehyde, terephthaldialdehyde, terephthaladehyde, terephthaldicarboxaldehyde, terephthalic aldehyde, 1,4-phthalaldehyde, p-formylbenzaldehyde, terephtaldehyde | |
| terephthalaldehyde |
Safety and Handling
EINECSNumber : (3)-3622
RTECSNumber : WZ0430000
TSCA : Yes