missing translation for 'onlineSavingsMsg'
Learn More

Terephthalaldehyde 98.0+%, TCI America™

Catalog No. p-7136025
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
T001025G 25 g
T0010100G 100 g
T0010500G 500 g
3 options

Catalog No. T001025G

Supplier: TCI America T001025G

Only null left

Chemical Identifiers

CAS 623-27-8
Molecular Formula C8H6O2
Molecular Weight (g/mol) 134.134
MDL Number MFCD00006949
InChI Key KUCOHFSKRZZVRO-UHFFFAOYSA-N
Synonym 1,4-benzenedicarboxaldehyde, p-phthalaldehyde, terephthaldehyde, terephthaldialdehyde, terephthaladehyde, terephthaldicarboxaldehyde, terephthalic aldehyde, 1,4-phthalaldehyde, p-formylbenzaldehyde, terephtaldehyde
PubChem CID 12173
IUPAC Name terephthalaldehyde
SMILES C1=CC(=CC=C1C=O)C=O

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 25 g
Formula Weight 134.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Terephthalaldehyde
EINECSNumber (3)-3622
RTECSNumber WZ0430000
TSCA Yes