missing translation for 'onlineSavingsMsg'
Learn More

Terephthalaldehyde mono(diethyl acetal), 97%, stab.

Catalog No. p-7022573
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1541306 5 g
AAA1541314 25 g
AAA1541322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1541306 Supplier Thermo Scientific Chemicals Supplier No. A1541306
Only null left

CAS: 81172-89-6 | C12H16O3 | 208.26 g/mol

4-(Diethoxymethyl)benzaldehyde was used in the synthesis of 4-(diethoxymethyl)benzyl alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Diethoxymethyl)benzaldehyde was used in the synthesis of 4-(diethoxymethyl)benzyl alcohol.

Solubility
soluble in alcohol, chlorinated solvents and toluene. Decomposes in water.

Notes
Air sensitive. Store under inert gas. Keep way from oxidizing agents, air.

Chemical Identifiers

CAS 81172-89-6
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.26
MDL Number MFCD00010217
InChI Key HTMXMFARWHNJDW-UHFFFAOYSA-N
Synonym 4-diethoxymethyl benzaldehyde, terephthalaldehyde mono diethyl acetal, benzaldehyde, 4-diethoxymethyl, terephthaldehyde mono diethyl acetal, terephthalaldehyde mono-diethyl acetal, pubchem22621, acmc-209tbs, 4-diethoxymethyl-benzaldehyde, 4-diethoxymethy benzaldehyde, 4-diethyoxymethyl benzaldehyde
PubChem CID 595993
IUPAC Name 4-(diethoxymethyl)benzaldehyde
SMILES CCOC(OCC)C1=CC=C(C=O)C=C1

Specifications

Density 1.047
Boiling Point 89°C to 93°C (7 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.507
Quantity 5 g
Beilstein 4293872
Sensitivity Air Sensitive
Solubility Information soluble in alcohol,chlorinated solvents and toluene. Decomposes in water.
Formula Weight 208.26
Percent Purity 98%
Chemical Name or Material Terephthalaldehyde mono(diethyl acetal), stabilized
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only