missing translation for 'onlineSavingsMsg'
Learn More

Terbium ICP Standard, 1000 ppm Tb in 3% HNO3, Ricca Chemical

Catalog No. PTB1KN100
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
PTB1KN100 Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PTB1KN100 Supplier Ricca Chemical Company Supplier No. PTB1KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12037-01-3 , 7697-37-2 , 12037-01-3

Specifications

Packaging Natural LDPE Bottle
CAS Min % 93.0, 4.42, 0.11
CAS Max % 97.0, 4.6, 0.11
Odor Odorless
Concentration or Composition (by Analyte or Components) 1mL = 1mg Tb (1, 000ppm Tb)Tb4O7 in 3% HNO3
Chemical Name or Material Terbium ICP Standard
ShelfLife 18 months