CAS RN 12037-01-3
IUPAC Name:
tetraoxotetraterboxane
Synonyms:
Terbium(III,IV) oxide|terbium(III,IV) oxide
Molecular Weight (g/mol):
747.69
Molecular Formula:
O7Tb4
InChi Key:
UWZVBPNIJCKCMC-UHFFFAOYSA-N
SMILES:
O=[Tb]O[Tb](=O)O[Tb](=O)O[Tb]=O
1
–
2
of
2
results
1
Terbium, plasma standard solution, Specpure™ Tb 1000μg/mL
CAS: 12037-01-3 Molecular Formula: O7Tb4 Molecular Weight (g/mol): 747.69 MDL Number: MFCD00083151 InChI Key: UWZVBPNIJCKCMC-UHFFFAOYSA-N IUPAC Name: tetraoxotetraterboxane SMILES: O=[Tb]O[Tb](=O)O[Tb](=O)O[Tb]=O
2
Terbium ICP Standard, 1000 ppm Tb in 3% HNO3, Ricca Chemical
CAS: 12037-01-3