missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenofovir disoproxil fumarate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 461250050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Tenofovir disoproxil fumarate | |
| Authentic | |
| C19H30N5O10P·C4H4O4 | |
| tenofovir disoproxil fumarate, viread, tenofovir df, unii-ott9j7900i, tenofovir disoproxil fumarate usan, 9-r-2-bis isopropoxycarbonyl oxy methoxy phosphinyl methoxy propyl adenine fumarate, 9-r-2-bis isopropoxycarbonyl oxy methoxy phosphinyl methoxy propyl adenine, fumarate, r-5-2-6-amino-9h-purin-9-yl-1-methylethoxy methyl-2,4,6,8-tetraoxa-5-phosphanonanedioic acid, bis 1-methylethyl ester, 5-oxide, e-2-butenedioate 1:1, 9-r-2-bis isopropoxycarbonyl oxy methoxy phosphinyl methoxy propyl adenine fumarate 1:1 | |
| VCMJCVGFSROFHV-WZGZYPNHSA-N | |
| [[(2R)-1-(6-aminopurin-9-yl)propan-2-yl]oxymethyl-(propan-2-yloxycarbonyloxymethoxy)phosphoryl]oxymethyl propan-2-yl carbonate;(E)-but-2-enedioic acid | |
| 6398764 | |
| 635.51 |
| 202138-50-9 | |
| 97.5% min. (HPLC) | |
| 5 g | |
| Solubility in water: >10 mg/mL. | |
| CC(C)OC(=O)OCOP(=O)(COC(C)CN1C=NC2=C1N=CN=C2N)OCOC(=O)OC(C)C.C(=CC(=O)O)C(=O)O | |
| 635.51 | |
| CHEBI:63718 | |
| 98% |
Chemical Identifiers
| 202138-50-9 | |
| 635.51 | |
| tenofovir disoproxil fumarate, viread, tenofovir df, unii-ott9j7900i, tenofovir disoproxil fumarate usan, 9-r-2-bis isopropoxycarbonyl oxy methoxy phosphinyl methoxy propyl adenine fumarate, 9-r-2-bis isopropoxycarbonyl oxy methoxy phosphinyl methoxy propyl adenine, fumarate, r-5-2-6-amino-9h-purin-9-yl-1-methylethoxy methyl-2,4,6,8-tetraoxa-5-phosphanonanedioic acid, bis 1-methylethyl ester, 5-oxide, e-2-butenedioate 1:1, 9-r-2-bis isopropoxycarbonyl oxy methoxy phosphinyl methoxy propyl adenine fumarate 1:1 | |
| CHEBI:63718 | |
| CC(C)OC(=O)OCOP(=O)(COC(C)CN1C=NC2=C1N=CN=C2N)OCOC(=O)OC(C)C.C(=CC(=O)O)C(=O)O |
| C19H30N5O10P·C4H4O4 | |
| VCMJCVGFSROFHV-WZGZYPNHSA-N | |
| 6398764 | |
| [[(2R)-1-(6-aminopurin-9-yl)propan-2-yl]oxymethyl-(propan-2-yloxycarbonyloxymethoxy)phosphoryl]oxymethyl propan-2-yl carbonate;(E)-but-2-enedioic acid |
RUO – Research Use Only