missing translation for 'onlineSavingsMsg'
Learn More

Tenofovir disoproxil fumarate, 98%, Thermo Scientific Chemicals

Catalog No. p-7077878
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461250010 1 g
AC461250050 5 g
AC461250250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC461250010 Supplier Thermo Scientific Chemicals Supplier No. 461250010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 202138-50-9
Molecular Formula C19H30N5O10P·C4H4O4
Molecular Weight (g/mol) 635.51
InChI Key VCMJCVGFSROFHV-WZGZYPNHSA-N
Synonym tenofovir disoproxil fumarate, viread, tenofovir df, unii-ott9j7900i, tenofovir disoproxil fumarate usan, 9-r-2-bis isopropoxycarbonyl oxy methoxy phosphinyl methoxy propyl adenine fumarate, 9-r-2-bis isopropoxycarbonyl oxy methoxy phosphinyl methoxy propyl adenine, fumarate, r-5-2-6-amino-9h-purin-9-yl-1-methylethoxy methyl-2,4,6,8-tetraoxa-5-phosphanonanedioic acid, bis 1-methylethyl ester, 5-oxide, e-2-butenedioate 1:1, 9-r-2-bis isopropoxycarbonyl oxy methoxy phosphinyl methoxy propyl adenine fumarate 1:1
PubChem CID 6398764
ChEBI CHEBI:63718
IUPAC Name [[(2R)-1-(6-aminopurin-9-yl)propan-2-yl]oxymethyl-(propan-2-yloxycarbonyloxymethoxy)phosphoryl]oxymethyl propan-2-yl carbonate;(E)-but-2-enedioic acid
SMILES CC(C)OC(=O)OCOP(=O)(COC(C)CN1C=NC2=C1N=CN=C2N)OCOC(=O)OC(C)C.C(=CC(=O)O)C(=O)O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: >10 mg/mL.
Formula Weight 635.51
Percent Purity 98%
Chemical Name or Material Tenofovir disoproxil fumarate

RUO – Research Use Only