accessibility menu, dialog, popup

UserName

CAS RN 202138-50-9

IUPAC Name: (2E)-but-2-enedioic acid; bis({[(propan-2-yloxy)carbonyl]oxy}methyl) {[(2R)-1-(6-amino-9H-purin-9-yl)propan-2-yl]oxy}methanephosphonate
Synonyms: viread tenofovir DF
Molecular Weight (g/mol): 635.52
Molecular Formula: C23H34N5O14P
InChi Key: VCMJCVGFSROFHV-WZGZYPNHSA-N
SMILES: OC(=O)\C=C\C(O)=O.CC(C)OC(=O)OCOP(=O)(CO[C@H](C)CN1C=NC2=C(N)N=CN=C12)OCOC(=O)OC(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results