missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Tenofovir

Catalog No. 366650R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
366650R 50 mg
366610R 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 366650R Supplier Tocris Bioscience™ Supplier No. 3666/50

Reverse transcriptase inhibitor

Tenofovir is a selectively inhibits HIV reverse transcriptase (RNA-dependent DNA polymerase). Prevents cytotoxicity in SIV-infected C-8166 cells in vitro (IC50 = 1.5 μM). Antiviral agent.

Specifications

CAS 147127-20-6
Quantity 50 mg
Molecular Formula (R)-[[2-(6-Amino-9 H-purin-9-yl)-1-methylethoxy]methyl]phosphonic acid
Product Type Tenofovir
Purity 99%
Molecular Weight (g/mol) M.W. (Observed): 287.21
Inhibitors Tenofovir

For Research Use Only