missing translation for 'onlineSavingsMsg'
Learn More

Teniposide 98.0+%, TCI America™

Catalog No. T310920MG
Change view
Click to view available options
Quantity:
20mg
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T310920MG 20mg
T3109100MG 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T310920MG Supplier TCI America Supplier No. T310920MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29767-20-2
Molecular Formula C32H32O13S

Specifications

Quantity 20mg
Melting Point 247°C
Formula Weight 656.66
Physical Form Crystalline Powder at 20°C
Color White
Purity ≥98.0% (HPLC)
Chemical Name or Material Teniposide
Recommended Storage Freezer (-20 °C)