missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Teniposide

Catalog No. AC466850010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466850010 1 g
AC466852500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466850010 Supplier Thermo Scientific Chemicals Supplier No. 466850010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29767-20-2
Molecular Formula C32H32O13S

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: insoluble.
Melting Point 242.0°C to 246.0°C
Formula Weight 656.66
InChI Key NRUKOCRGYNPUPR-UHFFFAOYNA-N
SMILES COC1=CC(=CC(OC)=C1O)C1C2C(COC2=O)C(OC2OC3COC(OC3C(O)C2O)C2=CC=CS2)C2=CC3=C(OCO3)C=C12
IUPAC Name 16-{[7,8-dihydroxy-2-(thiophen-2-yl)-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl]oxy}-10-(4-hydroxy-3,5-dimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one
Molecular Weight (g/mol) 656.66
Chemical Name or Material Teniposide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement May cause cancer.

Precautionary Statement GHS P Statement Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

EINECSNumber 249-831-2

RUO – Research Use Only