missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Tenax™ TA Glass Thermal Desorption Tubes

Catalog No. 111005376
Change view
Click to view available options
Matrix:
Tenax TA Porous Polymer (35/60)
Tenax TA Porous Polymer (60/80)
Packaging Quantity:
1/Pk.
10/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Matrix Packaging Quantity
111005376 Tenax TA Porous Polymer (60/80) 10/Pk.
111001618 Tenax TA Porous Polymer (35/60) 1/Pk.
111002484 Tenax TA Porous Polymer (60/80) 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111005376 Supplier MilliporeSigma™ Supelco™ Supplier No. 30134U

Suitable for use with Air Monitoring

  • Fritted
  • Thermally conditioned
  • Glass frit incorporated at the inlet for increased performance
  • Batch tested for back-pressure and background

Specifications

Overall Length (Metric) 88.9 mm
Includes 10 Thermal Desorption Tubes, PTFE Endcaps
Diameter (Metric) Outer 6.35 mm
Diameter (English) Outer 1/4 in.
Matrix Tenax TA Porous Polymer (60/80)
Methods EPA TO-1
For Use With (Application) Air Monitoring
Product Line Tenax
Description Unconditioned/Sealed with PTFE Endcaps
Type Thermal Desorption Tube
Packaging Quantity 10/Pk.
Overall Length (English) 3.5 in.
Material Glass
For Use With (Equipment) PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
Show More Show Less

Tenax is a registered trademark of Buchem B.V.