missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Temozolomide

Catalog No. AC466760010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466760010 1 g
AC466760050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466760010 Supplier Thermo Scientific Chemicals Supplier No. 466760010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 85622-93-1 | C6H6N6O2, C6H6N6O2 | 194.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85622-93-1
Molecular Formula C6H6N6O2

Specifications

Quantity 1 g
Formula Weight 194.15
InChI Key BPEGJWRSRHCHSN-UHFFFAOYSA-N
SMILES CN1N=NC2=C(N=CN2C1=O)C(N)=O
IUPAC Name 3-methyl-4-oxo-3H,4H-imidazo[4,3-d][1,2,3,5]tetrazine-8-carboxamide
Molecular Weight (g/mol) 194.15
Chemical Name or Material Temozolomide
What is the recommended solubility of Temozolomide?

The solubility of temozolomide is 10 mg/mL in DMSO, which is what we recommend for reconstitution.

Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Harmful if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

May cause cancer.

May damage fertility or the unborn child.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only