missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Telaprevir
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 1 g |
| Solubility | Solubility aq. soln.: .. Other solubilities: soluble in chloroform, methanol and dimethylsulfox, ide |
| Melting Point | 221.0°C to 224.0°C |
| Formula Weight | 679.86 |
| InChI Key | BBAWEDCPNXPBQM-IGSHNTALSA-N |
| SMILES | CCCC(NC(=O)[C@@H]1[C@H]2CCC[C@H]2CN1C(=O)[C@@H](NC(=O)[C@@H](NC(=O)C1=CN=CC=N1)C1CCCCC1)C(C)(C)C)C(=O)C(=O)NC1CC1 |
| IUPAC Name | 3-{[(1S,3aR,6aS)-2-[(2S)-2-[(2S)-2-cyclohexyl-2-[(pyrazin-2-yl)formamido]acetamido]-3,3-dimethylbutanoyl]-octahydrocyclopenta[c]pyrrol-1-yl]formamido}-N-cyclopropyl-2-oxohexanamide |
| Molecular Weight (g/mol) | 679.86 |
| Chemical Name or Material | Telaprevir |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling. |
RUO – Research Use Only