missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Telaprevir

Catalog No. AC466892500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466892500 250 mg
AC466890010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466892500 Supplier Thermo Scientific Chemicals Supplier No. 466892500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 402957-28-2 | C36H53N7O6, C36H53N7O6 | 679.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 402957-28-2
Molecular Formula C36H53N7O6

Specifications

Quantity 250 mg
Solubility Solubility aq. soln.: .. Other solubilities: soluble in chloroform, methanol and dimethylsulfox, ide
Melting Point 221.0°C to 224.0°C
Formula Weight 679.86
Chemical Name or Material Telaprevir
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

RUO – Research Use Only