missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Teen Prep Lysing Matrix SS, 15mL

Teen Prep Lysing Matrix SS is used for nucleic acid isolation and purification and to break hard/brittle samples

$68.96 - $335.98

Specifications

Content And Storage Room temperature
Sample Type Animal Tissue, Bacteria (Gram +/-), Bone, Feces, Fixed Samples, Food, Fungi, Heart, Lung, Plants, Roots, Seeds, Skeletal Muscle, Sludge, Soil, Spores, Wastewater, Yeast
For Use With (Equipment) FastPrep-24 5G, FastPrep-24, FastPrep-96
For Use With (Application) Nucleic acid isolation and purification
Matrix Stainless-Steel
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP116942005
View Documents Promotion Details
MP Biomedicals™
116942005
5 x 15 mL
Each for $68.96
Only null left
 
MP116942025
View Documents Promotion Details
MP Biomedicals™
116942025
25 x 15 mL
Each for $335.98
Only null left
 
Description

Description

  • Each impact-resistant 15mL tube contains fifteen 6.35 mm (0.25 in.) (0.25 cal) stainless-steel grinding ball
  • Specialty applications include RNA extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing

Certifications

DNase and RNase free

Specifications

Specifications

Room temperature
FastPrep-24 5G, FastPrep-24, FastPrep-96
Stainless-Steel
Animal Tissue, Bacteria (Gram +/-), Bone, Feces, Fixed Samples, Food, Fungi, Heart, Lung, Plants, Roots, Seeds, Skeletal Muscle, Sludge, Soil, Spores, Wastewater, Yeast
Nucleic acid isolation and purification
Lysis Matrix
Videos
SDS
Documents

Documents

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.