missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TDZD 8

Catalog No. 609250
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
609250 50 mg
609210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 609250 Supplier Tocris Bioscience™ Supplier No. 6092/50
Only null left

Selective non-ATP competitive inhibitor of GSK 3β

TDZD 8 is a selective non-ATP competitive inhibitor of GSK 3β (IC50 = 2 μM); thiadiazolidinone deriviative. Does not inhibit Cdk-1/cyclin B, CK-II, PKA or PKC at >100 μM. Reduces severity of L-dopa-induced dyskinesia in a Parkinson's disease in vivo model.

Specifications

CAS 327036-89-5
Quantity 50 mg
Solubility Soluble to 100mM in DMSO, 100mM in ethanol
Molecular Formula C10H10N2O2S
Formula Weight 222.26g/mol
Color White
Product Type Inhibitor

For Research Use Only