missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TDZD 8
Description
TDZD 8 is a selective non-ATP competitive inhibitor of GSK 3β (IC50 = 2 μM); thiadiazolidinone deriviative. Does not inhibit Cdk-1/cyclin B, CK-II, PKA or PKC at >100 μM. Reduces severity of L-dopa-induced dyskinesia in a Parkinson's disease in vivo model.
Specifications
Specifications
| CAS | 327036-89-5 |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO, 100mM in ethanol |
| Molecular Formula | C10H10N2O2S |
| Formula Weight | 222.26g/mol |
| Color | White |
| Product Type | Inhibitor |
For Research Use Only