missing translation for 'onlineSavingsMsg'
Learn More

Cupriethylenediamine, 1.0 M, Solution, Spectrum™ Chemical

Catalog No. 18601931
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601931 4 L Amber Glass Bottle
18601930 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18601931 Supplier Spectrum Chemical Mfg Cor Supplier No. C14494LTGL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

C1449, 14552-35-3, C4H18N4O2Cu

Spectrum™ Chemical Cupriethylenediamine, 1.0 M, Solution

Chemical Identifiers

CAS 13426-91-0 , 7732-18-5
Molecular Formula C4H16CuN4
Molecular Weight (g/mol) 183.75
InChI Key ATSGLBOJGVTHHC-UHFFFAOYSA-N
IUPAC Name copper(2+) bis(ethane-1,2-diamine)
SMILES [Cu++].NCCN.NCCN

Specifications

Color Purple
CAS Max % 22.5%, 77.5%
UN Number UN1761
Concentration 1.0 M
Grade Chemical Solution
Packaging Amber Glass Bottle
pH 12
Quantity 4 L
Chemical Name or Material Cupriethylenediamine