missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TCS 2002
Description
Potent inhibitor of GSK-3β (IC50 = 35 nM). Oral adminstration inhibits cold water stress-induced tau hyperphosphorylation in the mouse brain.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 338.38 |
| Purity | >99% |
| Molecular Weight (g/mol) | 338.381 |
| Chemical Name or Material | TCS 2002 |
Safety and Handling
| Recommended Storage | Store at +4°C |