missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCLP Standard 1 For Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$808.59 - $808.59
Chemical Identifiers
| CAS | 7697-37-2 |
|---|---|
| Molecular Formula | HNO3 |
| Molecular Weight (g/mol) | 63.01 |
| MDL Number | MFCD00011349 |
| InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
| PubChem CID | 944 |
| ChEBI | CHEBI:48107 |
| IUPAC Name | nitric acid |
| SMILES | O[N+]([O-])=O |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ba | 500μg/mL |
| Ag | 25μg/mL |
| As | 25μg/mL |
| Cr | 25μg/mL |
| Pb | 25μg/mL |
| Cd | 5μg/mL |
| Se | 5μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
| 7697-37-2 | |
| 63.01 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| CHEBI:48107 | |
| O[N+]([O-])=O |
| HNO3 | |
| MFCD00011349 | |
| 944 | |
| nitric acid |
Specifications
| Triple-leached LDPE bottle | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| MFCD00011349 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| 2% HNO3 | |
| 63.01 | |
| 944 | |
| Environmental | |
| TCLP Standard 1 |
| 7697-37-2 | |
| HNO3 | |
| AA, ICP | |
| Certified Reference Material | |
| O[N+]([O-])=O | |
| nitric acid | |
| 125 mL | |
| CHEBI:48107 | |
| Components in 5% HNO3: As •25μg/mL Ba •500μg/mL Cd •5μg/mL Cr •25μg/mL Pb •25μg/mL Hg† •100μg/mL Se •5μg/mL Ag •25μg/mL †Supplied separately. |
Safety and Handling
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only