missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TC-G 1004

Catalog No. 440750
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
440750 50 mg
440710 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 440750 Supplier Tocris Bioscience™ Supplier No. 4407/50

Potent and selective A2A antagonist

Potent antagonist of adenosine A2A receptors; displays >100-fold selectivity for A2A over A1 receptors (Ki values are 0.44 and 85 nM respectively). Potentiates L-DOPA-induced rotational behavior in 6-OHDA-lesioned rats.

Chemical Identifiers

CAS 1061747-72-5
Molecular Formula C22H27N7O2

Specifications

Quantity 50 mg
Formula Weight 421.5
Purity >98%
Molecular Weight (g/mol) 421.505
Chemical Name or Material TC-G 1004
Recommended Storage Store at -20°C