missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ TC ASK 10

Catalog No. 482550
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
482550 50 mg
482510 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 482550 Supplier R&D Systems™ Supplier No. 4825/50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Potent and selective ASK1 inhibitor; orally bioavailable

Potent ASK1 inhibitor (IC50 = 14 nM); displays selectivity for ASK1 over other kinases including ASK2 (IC50 = 0.51 μM), MEKK1, TAK1, IKKβ, ERK1, JNK1, p38α, GSK-3β, PKCθ and B-raf (IC50 values are > 10 μM). Blocks downstream JNK1/p38 phosphorylation in cells. Orally bioavailable.

Chemical Identifiers

CAS 1005775-56-3
Molecular Formula C21H23Cl2N5O

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 432.35
Purity >98%
Molecular Weight (g/mol) 432.349
Chemical Name or Material TC ASK 10
Recommended Storage Desiccate at Room Temperature