missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology, Fisher BioReagents™
Description
TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology
- Save time and simplify your buffer preparation step by using concentrated Fisher BioReagents 10X TBS buffer solution - simply dilute as needed
- 10X solution contains 1.37M NaCl, 0.027M KCl and 0.25M tris
- To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TBS solution with 9 volumes of ultrapure water, such as BP2819
- Filtered and autoclaved - ready to use stock TBS buffer
- Tested for the absence of DNase, RNase, and protease
- TBS buffer is often used in immunodetection techniques such as Western blotting protocols
- Intended for laboratory research use only
Specifications
Specifications
| Chemical Name or Material | Tris Buffered Saline |
| Name Note | 10X Solution |
| CAS | 7447-40-7, 7647-01-0, 7647-14-5, 77-86-1, 7732-18-5 |
| Boiling Point | 100°C |
| Color | Colorless |
| pH | 7.5 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| DNase | DNase free |
| Purity Grade Notes | DNase-, RNase- and Protease-Free |
| Show More |