missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ TBE Buffer, 10X, Molecular Biology Grade, Calbiochem™,

Catalog No. 5747951L
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5747951L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5747951L Supplier MilliporeSigma™ Supplier No. 5747951L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A 10X concentrate that can be diluted to a 1X solution containing 89mM Tris, 89mM boric acid, 2mM EDTA

Specifications

Chemical Name or Material TBE Buffer
CAS 10043-35-3
For Use With (Application) Electrophoresis
pH 8.3
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
DNase None Detected
Protease None Detected
Synonym Tris-Borate-EDTA Buffer
Grade Molecular Biology
Concentration or Composition (by Analyte or Components) 1X solution contains 89mM Tris, 89mM boric acid, 2mM EDTA
Show More Show Less