missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tazemetostat, 95%

Catalog No. AC468171000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468171000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC468171000 Supplier Thermo Scientific™ Supplier No. 468171000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Tazemetostat
CAS 1403254-99-8
Synonym EPZ-6438
Quantity 100 mg
Molecular Formula C34 H44 N4 O4
Formula Weight 572.75
Physical Form Solid
Product Type Reagent
Purity 95%