missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Tazarotene | |
| 105°C | |
| C21H21NO2S | |
| 100 mg | |
| OGQICQVSFDPSEI-UHFFFAOYSA-N | |
| ethyl 6-[2-(4,4-dimethyl-2,3-dihydrothiochromen-6-yl)ethynyl]pyridine-3-carboxylate | |
| 5381 | |
| 351.46 | |
| Crystalline Powder |
| 118292-40-3 | |
| Yellow | |
| MFCD00867628 | |
| tazarotene, tazorac, zorac, avage, ethyl 6-4,4-dimethylthiochroman-6-yl ethynyl nicotinate, fabior, tazarotene usan:inn, tazarotene avage, tazaroteno, tazarotenum | |
| CCOC(=O)C1=CN=C(C=C1)C#CC2=CC3=C(C=C2)SCCC3(C)C | |
| 351.464 | |
| CHEBI:32184 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 118292-40-3 | |
| 351.464 | |
| OGQICQVSFDPSEI-UHFFFAOYSA-N | |
| 5381 | |
| ethyl 6-[2-(4,4-dimethyl-2,3-dihydrothiochromen-6-yl)ethynyl]pyridine-3-carboxylate |
| C21H21NO2S | |
| MFCD00867628 | |
| tazarotene, tazorac, zorac, avage, ethyl 6-4,4-dimethylthiochroman-6-yl ethynyl nicotinate, fabior, tazarotene usan:inn, tazarotene avage, tazaroteno, tazarotenum | |
| CHEBI:32184 | |
| CCOC(=O)C1=CN=C(C=C1)C#CC2=CC3=C(C=C2)SCCC3(C)C |
Safety and Handling
RTECSNumber : US5675100
TSCA : No
Recommended Storage : Refrigerator