missing translation for 'onlineSavingsMsg'
Learn More

Tazarotene 98.0+%, TCI America™

Catalog No. p-7137579
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T310810MG 10 mg
T3108100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T310810MG Supplier TCI America Supplier No. T310810MG

Chemical Identifiers

CAS 118292-40-3
Molecular Formula C21H21NO2S
Molecular Weight (g/mol) 351.464
MDL Number MFCD00867628
InChI Key OGQICQVSFDPSEI-UHFFFAOYSA-N
Synonym tazarotene, tazorac, zorac, avage, ethyl 6-4,4-dimethylthiochroman-6-yl ethynyl nicotinate, fabior, tazarotene usan:inn, tazarotene avage, tazaroteno, tazarotenum
PubChem CID 5381
ChEBI CHEBI:32184
IUPAC Name ethyl 6-[2-(4,4-dimethyl-2,3-dihydrothiochromen-6-yl)ethynyl]pyridine-3-carboxylate
SMILES CCOC(=O)C1=CN=C(C=C1)C#CC2=CC3=C(C=C2)SCCC3(C)C

Specifications

Melting Point 105°C
Color Yellow
Quantity 10 mg
Formula Weight 351.46
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tazarotene
RTECSNumber US5675100
TSCA No
Recommended Storage Refrigerator