missing translation for 'onlineSavingsMsg'
Learn More

Tartaric Acid, Certified, 10.0% (w/v) ±0.5% (w/v), LabChem™

Catalog No. LC259702
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC259702 1 L Poly Bottle
LC259701 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC259702 Supplier LabChem Supplier No. LC259702

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87-69-4 , 7732-18-5
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
PubChem CID 444305
ChEBI CHEBI:15671
IUPAC Name 2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

Color Colorless to Yellow
CAS Max % 90.4, 9.6
Density 1.04g/mL
Quantity 1 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.04g/mL
Linear Formula C4H6O6
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 150.09
Percent Purity 10.0% ±0.5% (w/v)
Grade Certified
Chemical Name or Material Tartaric Acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear eye protection.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Recommended Storage Refrigerate