missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqPath™ 1-Step RT-qPCR Master Mix, CG

Catalog No. 50612244
Encompass
Change view
Click to view available options
Quantity:
5 x 1 mL
1 x 10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50612244 5 x 1 mL
501148245 1 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50612244 Supplier Applied Biosystems™ Supplier No. A15299

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TaqPath 1-Step RT-qPCR Master Mix, CG, is designed for robust and reproducible one-step pathogen detection and gene expression workflows. The single-tube 4X formulation contains thermostable MMLV reverse transcriptase, dNTPs, UNG, ROX dye, and thermostable Fast DNA polymerase.

TaqPath 1-Step RT-qPCR Master Mix, CG, is designed for robust and reproducible one-step pathogen detection and gene expression workflows. The single-tube 4X formulation contains thermostable MMLV reverse transcriptase, dNTPs, UNG, ROX dye, and thermostable Fast DNA polymerase, facilitating easy reaction set up—just add user-supplied assay and sample (Figure 1). All manufacturing lots are functionally tested to ensure remarkable lot-to-lot reproducibility for CT consistency and dynamic range across a wide variety of assays. TaqPath 1-Step RT-qPCR Master Mix, CG, is a General Purpose Reagent (GPR) is labeled 'For Laboratory Use' and, manufactured in an ISO 13485-certified and FDA-registered facility that adheres to cGMP principles. With stringent quality coupled with premier performance, TaqPath 1-Step RT-qPCR Master Mix, CG, is a superior choice for your diagnostic or biopharmaceutical testing and development needs.

Features of the TaqPath 1-Step RT-qPCR Master Mix, CG, include:

  • High sensitivity to detect low-copy targets with reproducible CT results
  • 6 logs of dynamic range with RNA, DNA, and multiplexing applications
  • Tolerance of inhibitors commonly found in clinical and biopharma samples
  • Advanced manufacturing production and process controls ensure lot-to-lot consistency
  • Compliance Package containing most frequently requested compliance documents, available below, commonly used to comply with internal quality systems and regulatory requirements in molecular diagnostic assay and drug development

High Sensitivity

With understanding of the importance of reproducible detection for low-titer pathogen or transcripts in clinical diagnostic testing, bioprocess QC testing, and adventitious testing, TaqPath 1-Step RT-qPCR Master Mix, CG, has been optimized as a higher-concentration 4X master mix that allows you to input more sample into each reaction, increasing sensitivity even in low-volume reactions. Figure 2 shows consistent CT results obtained from three unique lots when detecting 10 copy inputs of RNA target. Figure 3 demonstrates CT lot-to-lot consistency is preserved across multiple assays with different attributes and expression levels.

Wide Dynamic Range Compatible with RNA, DNA, and Multiplexing

TaqPath 1-Step RT-qPCR Master Mix, CG, has been optimized to provide high specificity and dynamic range for use with both RNA and DNA targets. Since virology labs often test for both RNA and DNA viruses, TaqPath 1-Step RT-qPCR Master Mix, CG, is designed to use a single protocol to assay both types of nucleic acid. This input flexibility can help streamline the number of different workflows in your lab to improve efficiency. Figure 4 demonstrates the excellent PCR linearity across a 6-log input range for both RNA (Figure 4a) and DNA targets (Figure 4b).

TaqPath 1-Step RT-qPCR Master, CG, is compatible with multiplexing reactions, allowing for additional exogenous or endogenous controls or targets to be run simultaneously for quality control or efficiency purposes. Figure 5 demonstrates TaqPath 1-Step RT-qPCR Master Mix performance in a triplex reaction. The formulation can be run in either fast or standard cycling conditions with equivalent performance across a wide variety of qPCR platforms.

TaqPath 1-Step RT-qPCR Master Mix, CG, has been benchmarked against similar competitor master mixes and demonstrates equivalent or better sensitivity and dynamic range across a variety of targets (Figure 6).

Inhibitor Tolerant

Unlike other master mixes on the market, TaqPath 1-Step RT-qPCR Master Mix's unique proprietary formulation allows robust performance even in the presence of PCR inhibitors typically found in biopharmaceutical, molecular diagnostics, and research applications. Optimized performance in the presence of such substances that can normally inhibit PCR, such as heparin, hematin, or EDTA, helps increase confidence when working with a variety of complex clinical or biopharma samples. TaqPath master mixes demonstrate increased tolerance of inhibitors compared to competitor mixes (Figure 7).

Manufacturing Production and Process Controls

TaqPath qPCR Master Mix is manufactured under a strong quality management system that utilizes traceable quality raw materials and validated operating procedures. With established controls from purchasing through QC release, TaqPath product manufacturing is designed to produce consistent product performance lot after lot. The stringent QC is important for a variety of applications, making the master mix ideal for bioprocess quality control testing, biologics adventitious testing, molecular diagnostics, and research applications.

General Purpose Reagent manufactured under cGMP

TaqPath master mixes are general purpose reagents labeled 'For Laboratory Use' and manufactured in an ISO 13485-certified and FDA-registered facility that adheres to cGMP principles.

Specifications

Concentration 4X
Content And Storage Contains 5 x 1 mL tubes of 4X RT-PCR master mix sufficient for 1000 x 20 μL reactions.

Store at -15°C to -25°C in the dark.
Detection Method Primer-probe
Form Liquid
Format Tube
GC-Rich PCR Performance High
PCR Method 1-step RT-qPCR
Polymerase Fast DNA Polymerase
Reaction Speed Fast or Standard
For Use With (Equipment) 7500 Fast System, 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, StepOnePlus™, ViiA™ 7 System, 7500 Fast Dx System, QuantStudio™ Dx
Multiplex Capability Up to 3-plex
Passive Reference Dye ROX (Pre-mixed)
Product Line TaqPath
Product Type RT-qPCR Master Mix
Quantity 5 x 1 mL
Reverse Transcriptase M-MLV
Sensitivity Down to 10 copies
Sufficient For 1000 Reactions
Target Specificity RNA only, Both RNA and DNA
For Use With (Application) Microbial Detection, DNA Quantification, Presence/Absence
Thermostability Not Applicable
Fidelity (vs. Taq) 4 X
Hot Start Built-In Hot Start
No. of Reactions 1000 Reactions
Sample Type RNA, cDNA, DNA
Volume 1 mL
Includes dUTP, UNG/UDG
Inhibitor Tolerance (Simple) High
Show More Show Less
What is the recommended RNA input range for TaqPath 1-Step RT-qPCR Master Mix, CG (Cat. Nos. A15299, A15300)?

The figure for TaqPath 1-Step RT-qPCR Master Mix, CG (Cat. Nos. A15299, A15300) below, shows the dynamic range of RNA Triplex Amplification at 0.1 pg–100 ng. While a reaction volume is not specified, we typically use 20 µL reactions.
RNA Triplex Amplification

What starting template can I use with the TaqPath 1-Step RT-qPCR Master Mix?

You can use this protocol for both RNA and DNA targets. During thermal cycling, the reverse transcription step will not affect performance with DNA targets.

For Laboratory Use.