missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Arbovirus Triplex Control Kit (ZIKV/DENV/CHIKV)

Catalog No. A40490
Change view
Click to view available options
No. of Reactions:
25 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
A40490 25 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A40490 Supplier Applied Biosystems™ Supplier No. A40490

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Applied Biosystems™ TaqMan Zika Virus Triplex Control Kit (ZIKV/DENV/CHIKV) includes a single foil pouch that contains human DNA at 1X and pooled complete genome RNA template of multiple viruses.

The Applied Biosystems™ TaqMan Zika Virus Triplex Control Kit (ZIKV/DENV/CHIKV) includes a single foil pouch that contains human DNA at 1X and pooled complete genome RNA template of the following viruses:

  • Zika virus with Asian lineage
  • Dengue virus from 4 serotypes (DENV1, DENV2, DENV3, and DENV4)
  • Chikungunya virus

 The control kit is used with the following kits:

  • TaqMan Zika Virus Triplex Kit (ZIKV/DENV/CHIKV) (0.2 mL block)
  • TaqMan Zika Virus Triplex Kit (ZIKV/DENV/CHIKV) (0.1 mL block)

Specifications

For Use With TaqMan™ Zika Virus Triplex Kit
Product Line TaqMan
Shipping Condition Room Temperature
No. of Reactions 25 Reactions