missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Urinary Tract Microbiota Amplification Control

Catalog No. A39174
Encompass
Change view
Click to view available options
Quantity:
5 x 50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A39174 5 x 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A39174 Supplier Applied Biosystems™ Supplier No. A39174

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The TaqMan Urinary Tract Microbiota Amplification Control is designed to evaluate the amplification efficacy of the TaqMan Urinary Tract Microbiota assays.

It is a multi-target plasmid that includes amplicons for each of the qualified TaqMan Urinary Tract Microbiota assays, as well as control prokaryotic 16S rRNA and human RNase P RPPH1 gene sequences. This plasmid serves as a positive control for the TaqMan Urinary Tract Microbiota Assay collection.

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Format Tube(s)
Product Line TaqMan
Product Type Urinary Tract Microbiota Amplification Control
Quantity 5 x 50 μL
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Species E. coli
Does the TaqMan Urinary Tract Microbiota Amplification Control (Cat. No. A39174) contain 16S rRNA and RNase P?

The TaqMan Urinary Tract Microbiota Amplification Control (Cat. No. A39174) contains a linearized plasmid harboring multi-target sequences, including the sequences of 16S rRNA and the RNase P genes. There is no separate 16S or RNase P control in this kit.

Which sample prep kit do you recommend when running urinary tract microbiota profiling experiments?

We recommend the MagMAX DNA Multi-Sample Ultra Kit (Cat. No. A25597) for isolating DNA for urinary tract microbiota profiling experiments.

Do you have any positive controls for the urinary tract microbiota assays?

The TaqMan Urinary Tract Microbiota Amplification Control (Cat. No. A39174) is a linearized multi-target plasmid with target sequences for each available urinary tract microbiota profiling assay.

How can I check for the presence of inhibitors in my samples when running microbiota profiling experiments?

We offer a TaqMan Universal DNA Spike-In Control (Cat. No. A39175 or A39176) that can be added to samples before extraction, to monitor recovery efficiency.

For Research Use Only. Not for use in diagnostic procedures.