missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Universal DNA Spike-In Control (5X)

Catalog No. A39176
Encompass
Change view
Click to view available options
Quantity:
5 x 1000 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A39176 5 x 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A39176 Supplier Applied Biosystems™ Supplier No. A39176

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TaqMan Universal DNA Spike-In Control serves as an exogenous process control of a known concentration that is spiked into a sample at specific stages throughout the qPCR process.

Measuring the concentration of the spiked-in sequence allows for its use as a serial quality control and it can be used to monitor recovery efficiency and to identify false negative results such as the presence of PCR inhibitors in molecular detection workflows. This is of particular importance when working with sample types that have a high level of inhibition. For a 1X version of this control, please see Cat. No. A39175.

Specifications

Concentration 5X
Content And Storage Store in freezer (-5°C to -30°C).
Format Tube(s)
GC-Rich PCR Performance High
PCR Method qPCR
Product Line TaqMan
Product Type Universal DNA Spike-In Control
Quantity 5 x 1000 μL
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Species Not Species-Specific
Fidelity (vs. Taq) 5 X
Show More Show Less
How can I check for the presence of inhibitors in my samples when running microbiota profiling experiments?

We offer a TaqMan Universal DNA Spike-In Control (Cat. No. A39175 or A39176) that can be added to samples before extraction, to monitor recovery efficiency.

For Research Use Only. Not for use in diagnostic procedures.