Learn More
Applied Biosystems™ TaqMan™ RNA-to-CT™ 1-Step Kit
Description
The TaqMan™ RNA-to-CT™ One-Step Kit is recommended for a variety of applications including general gene expression studies, biomarker discovery, and microarray validation.
- Accurate across a wide dynamic range for dependable performance
- High sensitivity, enabling low copy number detection and precise quantification
- Validated with TaqMan™ Gene Expression Assays for easy experimental setup
- Consistent performance with a wide variety of targets, including AT-rich, GC-rich and long sequences
- Fewer pipetting steps means less hands-on-time and less chance for errors
Efficiency across a Broad Range of Input Concentrations
- Maximum flexibility in RNA template input quantity requires optimal real-time PCR efficiency across a broad range of template quantities
- The result is comparable to that of the TaqMan™ RNA-to-CT™ 2-Step Kit, and is a significant improvement in dynamic range and sensitivity over the TaqMan™ One-Step RT-PCR Master Mix Reagents.
High Sensitivity at Low Target Concentration
- The sensitivity of TaqMan™ RNA-to-CT™ One-Step Kit was validated using a synthetic RNA template for which copy number is precisely known
- Significant sampling error occurs when measuring low quantities of target, so statistical analysis is required for proper evaluation using multiple replicates
- Statistical analysis indicates high confidence of sample quantification based on a T-test, consistent with as few as 10 copies of target.
Order Info
Shipping Conditions: Wet Ice
Specifications
Specifications
| Content And Storage | Contains sufficient reagents for 200 reactions at 50 μl reaction volumes. Includes: • 1 × 5 ml tube of 2X master mix containing AmpliTaq Gold™ DNA Polymerase UP, dNTPs (including dUTP), Passive Reference 1, and optimized buffer components. Store at -15 to -25°C. • 1 × 250 μl tube of 40X RT enzyme mix containing ArrayScript™ UP Reverse Transcriptase, RNase Inhibitor. Store at -15°C to -30°C. |
| Detection Method | Primer-probe |
| PCR Method | 1-step RT-qPCR |
| Polymerase | AmpliTaq Gold DNA Polymerase |
| Reaction Speed | Standard |
| For Use With (Equipment) | 7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode |
| Passive Reference Dye | ROX (Pre-mixed) |
| Product Line | RNA-to-CT, TaqMan |
| Product Type | One-Step RT-PCR Kit |
| Purity or Quality Grade | UP (Ultra Pure) |
| Show More |
Frequently Asked Questions (FAQs)
If you are targeting a low-abundance gene, you may have trouble getting Ct values in a good, reliable range (Ct > 32). To increase the sensitivity of the assay, you may want to consider the following:
- Increase the amount of RNA input into your reverse transcription reaction, if possible
- Increase the amount of cDNA in your qPCR reaction (20% by volume max)
- Try a different reverse transcription kit, such as our SuperScript VILO Master Mix, for the highest cDNA yield possible
- Consider trying a one-step or Cells-to-CT type workflow (depending on your sample type)
Most times your instrument software can automatically set a proper baseline for your data. Check out our short video, Understanding Baselines, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=5BjFAJHW-bE).
In most cases your instrument software can automatically set a proper threshold for your data. Check out our short video, Understanding Thresholds, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=H_xsuRQIM9M).
There could be several reasons for no amplification from an assay or primer set. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/no-amplification.html) for more details.
There could be several reasons for amplification in a NTC well. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/amplification-no-template-control.html) for more details.
For Research Use Only. Not for use in diagnostic procedures.